Learn More
Medchemexpress LLC SU-4942 | 76086-99-2 | 99.6% | 300.15 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY2129010MM/1ML
Encompass_Preferred
SU-4942 is a tyrosine kinase signal modulator that inhibits VEGF- and endothelial cell growth factor (ECGF)-induced mitogenesis in endothelial cells.
  • Tyrosine kinase signal modulator.
  • Inhibits VEGF- and endothelial cell growth factor (ECGF)-induced mitogenesis in endothelial cells.
  • For research use only.
Catalog No. 50-004-03207