Learn More
Medchemexpress LLC SU-4942 | 76086-99-2 | 99.6% | 300.15 | 25 MG

Supplier: Medchemexpress LLC 25MGHY21290
SU-4942 is a tyrosine kinase signal modulator that inhibits VEGF- and endothelial cell growth factor (ECGF)-induced mitogenesis in endothelial cells. It is for research use only.
- Inhibits VEGF- and ECGF-induced mitogenesis
- Suitable for laboratory research
- High purity (99.60%)
- Available in various solid and solution forms
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.