Learn More
Medchemexpress LLC SU-4942 tyrosine kinase signal modulator | 76086-99-2 | 99.6% | 300.15 | 100 MG
Supplier: Medchemexpress LLC HY21290100MG
SU-4942 is a tyrosine kinase signal modulator that inhibits VEGF- and endothelial cell growth factor (ECGF)-induced mitogenesis in endothelial cells. This compound is suitable for various laboratory research applications.
- Functions as a tyrosine kinase signal modulator.
- Inhibits VEGF-induced mitogenesis.
- Inhibits ECGF-induced mitogenesis.