Learn More
Medchemexpress LLC Su 5402 | 215543-92-3 | MFCD08235144 | >95.0% | 296.32 | C17H16N2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1040750MG
Encompass_Preferred
SU 5402 is a multi-targeted receptor tyrosine kinase inhibitor active against VEGFR2, FGFR1, and PDGFRβ. It shows low-nanomolar potency for VEGFR2 and FGFR1, is supplied as a powder for research use, and has documented solubility and storage recommendations for experimental workflows.
  • Potent inhibition of VEGFR2, FGFR1, and PDGFRβ.
  • Low-nanomolar IC50 values for primary targets.
  • Suitable for in vitro and in vivo research applications.
  • Available in small milligram package sizes for laboratory use.
  • High solubility in DMSO and formulated solvent mixes.
  • Long-term stability when stored as powder at recommended temperatures.
Catalog No. 50-002-42203