Learn More
Medchemexpress LLC Su 5402 | 215543-92-3 | MFCD08235144 | 99.9% | 296.32 g·mol⁻¹ | C17H16N2O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1040725MG
Encompass_Preferred
SU 5402 is a small-molecule, multi-target receptor tyrosine kinase inhibitor for research use. It potently inhibits VEGFR2, FGFR1, and PDGFRβ and is supplied as a high-purity solid for in vitro and in vivo studies.
  • Inhibits VEGFR2 (IC50 20 nM), FGFR1 (IC50 30 nM), and PDGFRβ (IC50 510 nM).
  • High purity (>99.8%).
  • Molecular weight 296.32 g·mol⁻¹ and formula C17H16N2O3.
  • Supplied as a solid suitable for cell-based assays and animal studies.
  • ATP-competitive, reversible kinase inhibitor mechanism.
Catalog No. 50-002-42204