Learn More
Apexbio Technology LLC SU 5402, 25mg CAS# 215543-92-3
SDP
Supplier:  Apexbio Technology LLC A384325
Encompass_Preferred
IC50: 0.02, 0.03, 0.51 and > 100 M for VEGFR2, FGFR1, PDGFR and EGFR, respectivelySU 5402 is an inhibitor of VEGF, FGF, and PDGF receptor tyrosine kinases. Receptor tyrosine kinases have been recognized as therapeutic targets for the treatment of human diseases including cancers, inflammatory diseases, and cardiovascular diseases. In vitro: SU5402 could inhibit FGFR3 phosphorylation in vitro. B cells dependent on FGFR3 for survival were sensitive to SU5402 specifically. A panel of 11 human myeloma cell lines was studied, among which five beared t(4;14) translocation. The KMS11 human myeloma cell line expressesing constitutively active mutant FGFR3, displayed a 95% increase in G0/G1 cells as well as 45-fold increase in apoptotic cells after SU5402 treatment. In addition, the activated signal-regulated kinases 1 and 2 and signal activator of transcription 3 were down-regulated after SU5402 treatment rapidly. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8377