Learn More
Abcam SU-5402 (DMSO solution), VEGFR and FGFR inhibitor, 500UG-LQD
Supplier:  Abcam AB146602500UGLQD
Encompass_Preferred
MW 296.32 Da, Purity >98%. Potent, selective VEGFR and FGFR inhibitor (IC₅₀ values are 0.02 (VEGFR2), 0.03 (FGFR1), 0.51 (PDGFRβ) and >100 μM (EGFR)). Attenuates integrin β4-induced differentiation of neural stem cells. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4555