Learn More
Abcam SU 5416 (Sema x anib), VEGFR-2 inhibitor, 10MG
Supplier:  Abcam AB14505610MG
Encompass_Preferred
MW 238.28 Da, Purity >99%. Selective VEGFR-2 inhibitor. Adenine mimetic. Inhibits endothelial cell proliferation. Inhibits angiogenesis and shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5441