Learn More
KAYAKU ADVANCED MATERIALS INC SU-8 3025, negative epoxy-based photoresist, 500 mL, high chemical resistance, for microfabrication applications, imaging wavelength: near UV (350-400 nm), viscosity 4400 cP, density 1.143 g/mL
Supplier:  KAYAKU ADVANCED MATERIALS INC Y311072 0500L1GL
Encompass
SU-8 3025, negative epoxy-based photoresist, 500 mL, high chemical resistance, for microfabrication applications, imaging wavelength: near UV (350-400 nm), viscosity 4400 cP, density 1.143 g/mL
Catalog No. NC0057282