Learn More
Foxx Life Sciences EZBio MTO, Single Use (SUT) Bottle Assembly, 125mL Sterilized PC Bottle for Aseptic Sampling and Sterile Storage, 38-430, Flexelene®, Vented w/ Diptube, Luer, 10/CS
Supplier:  Foxx Life Sciences 4384203FLS
Encompass_Preferred
Made To Order MTO Single Use Sterile 125mL Polycarbonate Media Bottle Assemblies with Tubing and Vent Filters. EZBio MTO Single Use Media Bottle Assemblies include a 2-port 38-430 VersaCap assembly with 1/8" ID (3.2mm) x 1/4" OD (6.4mm) Flexelene® TPE Tubing and diptube. This bottle assembly features polypropylene luer locks that connect small diameter tubing with secure, leak-proof connections. Additionally, all bottle assemblies include a 0.2µm Hydrophobic PTFE Vent Filter that allows for ventilation while preventing media from escaping in the event of a spill. All EZBio "MTO" Single Use Media Bottle Assemblies also include an extra closed cap for storage and a ratchet style pinch clamp that allows the user to control flow into the container or close off the line entirely. The EZBio "MTO" Bottle Assembly is the perfect option to phase out costly stainless steel apparatuses that are expensive to clean and validate. USP Class VI components, Gamma Irradiated in USA.
Catalog No. 50-206-0461