Learn More
World Precision Instrument SU-M33A Micromanipulator-SU-MM3A-L
SDP
Supplier:  World Precision Instrument SUM33AL
Encompass
Successfully used in conjunction with our other micromanipulators, the SU-M33A is available separately for those in need of a small, practical instrument in applications not requiring submicron accuracy.
The SU-M33A is the right choice for tight environments which require maximum versatility within a small space. The vertical lock is ideal for locking the position of the vertical axis and increasing stability. The optional rotating base and tiltable X-axis provide even further flexibility.
Catalog No. 50-253-8540