Learn More
Medchemexpress LLC SU056 | 2376580-08-2 | C20H16FNO5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY150231100MG
Encompass_Preferred
SU056 is a YB-1 inhibitor. It induces cell-cycle arrest, apoptosis, and inhibits cell migration in ovarian cancer cells. It interacts with YB-1 and inhibits its associated downstream proteins and pathways. SU056 can enhance the cytotoxic effects of Paclitaxel.
  • Induces cell-cycle arrest, apoptosis, and inhibits cell migration in ovarian cancer cells.
  • Inhibits colony formation.
  • Inhibits tumor growth in vivo.
  • Enhances the cytotoxic effects of Paclitaxel.
Catalog No. 50-003-75310