Learn More
Apexbio Technology LLC SU11274 658084-23-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A26785.1
Encompass_Preferred
SU11274 (CAS 658084-23-2) is a selective inhibitor targeting Met kinase exhibiting an IC50 of approximately 10 nM The compound specifically suppresses the autophosphorylation of Met at residues Tyr1234/1235 disrupting receptor activity with minimal effects on other receptor tyrosine kinases such as EGFR and PDGFR or serine/threonine kinases including CDK2 In NIH3T3 cells expressing drug-sensitive or resistant MET mutants and in H1993 human lung cancer cells treatment with SU11274 induces a dose-dependent inhibition of Met phosphorylation and impairs receptor ubiquitination leading to receptor accumulation SU11274 serves as a useful tool for investigating Met signaling pathways in oncology research
Catalog No. 50-000-63557