Learn More
Selleck Chemical LLC SU11274 S1080-1g
SDP
Supplier:  Selleck Chemical LLC S10801G
Encompass_Preferred
SU11274 (PKI-SU11274) is a selective Met (c-Met) inhibitor with IC50 of 10 nM in cell-free assays no effects on PGDFR EGFR or Tie2 SU11274 induces autophagy apoptosis and cell cycle arrest
Catalog No. 50-310-8320