Learn More
Apexbio Technology LLC SU14813 627908-92-3 10mg
SDP
Supplier:  Apexbio Technology LLC A384410
Encompass_Preferred
SU14813 (CAS 627908-92-3) is a small molecule tyrosine kinase inhibitor designed for oral administration It inhibits multiple receptor tyrosine kinases including vascular endothelial growth factor receptors (VEGFR) platelet-derived growth factor receptors (PDGFR) c-Kit and fms-like tyrosine kinase 3 (FLT-3) Through the blockade of these signaling pathways SU14813 interferes with processes such as angiogenesis and tumor cell proliferation This compound is widely utilized in biomedical research to investigate mechanisms of tumor growth and neovascularization as well as to evaluate novel therapeutic strategies targeting angiogenic and oncogenic pathways
Catalog No. 50-000-96675