Learn More
Medchemexpress LLC Su14813 maleate | 849643-15-8 | MFCD24682713 | 99.0% | 558.56 g·mol⁻¹ | C27H31FN4O8 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10501A10MM 1ML
Encompass_Preferred
SU14813 maleate is a small-molecule, multi-targeted receptor tyrosine kinase inhibitor for research use. It potently inhibits VEGFR1, VEGFR2, PDGFRβ, and KIT in the low nanomolar range and is supplied as both a 10 mM solution in DMSO and as solid quantities for biochemical and cellular assays.
  • Inhibits VEGFR1, VEGFR2, PDGFRβ, and KIT with low-nanomolar IC50s.
  • Supplied as a 10 mM solution in DMSO and as solids for flexibility in experiments.
  • High reported purity suitable for research applications.
  • Molecular weight 558.56 g·mol⁻¹ and formula C27H31FN4O8 for reference.
  • Available in small quantities for assay development and dose-response studies.
  • Suitable for studying receptor tyrosine kinase signaling in vitro.
Catalog No. 50-002-43189