Learn More
Medchemexpress LLC SU1498 | 168835-82-3 | 98.8% | 390.52 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19326
Encompass_Preferred
SU1498 (AG 1498) is a selective inhibitor of the VEGFR2; it inhibits Flk-1 with an IC50 value of 700 nM.
  • Selective inhibitor of VEGFR2.
  • Inhibits Flk-1 with an IC50 of 700 nM.
Catalog No. 50-003-85588