Learn More
Abcam SU4312, VEGFR and PDGFR receptor tyrosine kinase inhibitor, 5MG
Supplier:  Abcam AB1415775MG
Encompass_Preferred
MW 264.32 Da, Purity >98%. Potent, selective VEGFR and PDGFR tyrosine kinases inhibitor (IC₅₀ values are 0.8 and 19.4 μM respectively). Shows anti-angiogenic effects. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6515