Learn More
Medchemexpress LLC Su4984 | 186610-89-9 | MFCD08443934 | 99.7% | 333.38 g/mol | C20H19N3O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY118203100MG
Encompass_Preferred
SU4984 is a small-molecule protein tyrosine kinase inhibitor that targets FGFR1 (reported IC50 10-20 μM), supplied as a 100 mg solid for laboratory research with a reported purity of 99.7%.
  • Inhibits FGFR1 with a reported IC50 of 10-20 μM.
  • Useful for research into cancer-related signaling pathways.
  • High purity (99.7%) suitable for biochemical and cellular assays.
  • Supplied as a 100 mg solid for convenient laboratory dosing.
  • Molecular weight 333.38 g/mol and chemical formula C20H19N3O2.
  • Provided with an SDS for safe handling and storage guidance.
Catalog No. 50-002-26825