Learn More
Medchemexpress LLC SU4984 | 186610-89-9 | MFCD08443934 | 99.7% | 333.38 g/mol | C20H19N3O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11820325MG
Encompass_Preferred
SU4984 is a small-molecule protein tyrosine kinase inhibitor that primarily targets fibroblast growth factor receptor 1 (FGFR1) with a reported IC50 of 10-20 μM. It also shows activity against platelet-derived growth factor receptor and the insulin receptor. Supplied for research use in a 25 mg vial with high purity.
Catalog No. 50-002-27739