Learn More
Medchemexpress LLC SU5201 | 114727-43-4 | 99.5% | 290.14 | 50 MG

Supplier: Medchemexpress LLC HY2129350MG
SU5201 is an inhibitor of interleukin-2 (IL-2) production. This compound is primarily targeted toward interleukin related pathways, specifically inhibiting IL-2. It appears as a solid, yellow to orange substance. For optimal stability, it requires storage at 4°C, protected from light.
- Inhibits interleukin-2 (IL-2) production
- Molecular formula is C15H9Cl2NO
- Molecular weight is 290.14
- Appears as a solid with a yellow to orange color
- Store at 4°C, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.