Learn More
Selleck Chemical LLC SU5204 S0290-5MG
SDP
Supplier:  Selleck Chemical LLC S02905MG
Encompass_Preferred
SU5204 is a tyrosine kinase inhibitor with IC50 of 4 M and 51 5 M for FLK-1 (VEGFR-2) and HER2 respectively
Catalog No. 50-000-43492