Learn More
Selleck Chemical LLC SU5208 S0291-5MG
SDP
Supplier:  Selleck Chemical LLC S02915MG
Encompass_Preferred
SU5208(3-[(Thien-2-yl)methylene]-2-indolinone) is an inhibitor of vascular endothelial growth factor receptor-2 (VEGFR2)
Catalog No. 50-000-43493