Learn More
Medchemexpress LLC Su5208 | 62540-08-3 | 99.5% | 227.28 g/mol | C13H9NOS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1362095MG
Encompass_Preferred
SU5208 is a small-molecule inhibitor of vascular endothelial growth factor receptor-2 (VEGFR2) supplied for laboratory research use. The compound (C13H9NOS; MW 227.28 g/mol) is available as a solid and as DMSO solutions and has reported activity in cellular assays.
  • Inhibits vascular endothelial growth factor receptor-2 (VEGFR2).
  • Reported cytotoxicity in A549 cells (IC50 reported).
  • Approximately 99.5% purity for research applications.
  • Available in multiple package sizes and as prepared DMSO solutions.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-23274