Learn More
Selleck Chemical LLC SU5402 10mg 215543-92-3
SDP
Supplier:  Selleck Chemical LLC S766710MG
Encompass_Preferred
SU5402 is a potent multi-targeted receptor tyrosine kinase inhibitor with IC50 of 20 nM, 30 nM, and 510 nM for VEGFR2, FGFR1, and PDGF-R?, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5101