Learn More
Selleck Chemical LLC SU6656 5mg 330161-87-0
SDP
Supplier:  Selleck Chemical LLC S77745MG
Encompass_Preferred
SU 6656 is a selective Src family kinase inhibitor with IC50 of 280 nM, 20 nM, 130 nM, and 170 nM for Src, Yes, Lyn, and Fyn, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8001