Learn More
Zen Bio Inc SUBCUTANEOUD HUMAN PREADIPOCYT
SDP
Supplier:  Zen Bio Inc SPFSL
Encompass
Cryopreserved Preadipocytes, frozen vial containing at least 2 x 106 Preadipocytes, 50mL Preadipocyte Medium, Store in liquid nitrogen, Storage Condition: Media - Short Term 4degreeC, 6 months -20degreeC
Catalog No. NC9994329