Learn More
Medchemexpress LLC Suberic acid (octanedioic acid) | 505-48-6 | MFCD00004428 | 98.0% | 174.20 | C8H14O4 | 25 G

Supplier: Medchemexpress LLC 25GHYW015300
Suberic acid (octanedioic acid) is a crystalline dicarboxylic acid supplied for research use as a reagent or analytical standard. It has reported biological activity (activates Akt; modulates TGF-β and MAPK pathways) and is offered as solid and as a ready solution for biochemical and signaling studies.
- Molecular formula C8H14O4.
- Molecular weight 174.20 g/mol.
- Purity 98.0%.
- Available as crystalline solid or 10 mM solution in DMSO.
- Pack sizes: 5 g, 10 g, 25 g, 50 g, 100 g, or larger on request.
- Intended for research and analytical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.