Learn More
Abcam Suberoyl bis-hydro x amic acid (SBHA), HDAC inhibitor, 100MG
Supplier:  Abcam AB144532100MG
Encompass_Preferred
MW 204.22 Da, Purity >98%. Potent, non-selective HDAC inhibitor (ID₅₀ values are 0.25 and 0.3 μM for HDAC1 and HDAC3 respectively). Potentiates the cytostatic effects of 5-Fluorouracil (ab142387). Induces apoptosis. Shows antiproliferative effects (IC₅₀ = 15 μM, HeLa cells).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5646