Learn More
Grainger Submersion pH Kit w/ATC Ryton PPS BNC
Supplier:  Grainger 16Z030
Encompass_Preferred
The SensoRex pH Submersion Sensor Kit is a high-quality pH measurement system designed for laboratory use. It features a glass pH electrode with built-in temperature sensor, BNC connector, and 3 m cable. The kit includes calibration and cleaning solutions, as well as a sturdy holder stand for submersion measurements. With an accuracy of ±0.02 pH, this professional-grade pH meter provides reliable results for critical laboratory applications. The SensoRex sensor is easy to use, maintain, and calibrate for consistent pH testing.
Catalog No. 50-276-8097