Learn More
Electron Microscopy Sciences MXS Calibration Standard MXS 701CE for SEM, Pin 700 nm, 700 nm Spacing

Supplier: Electron Microscopy Sciences 8011071PIN
EMS offers a series of calibration standards with one and two dimension calibrated patterns. These standards are created utilizing holographic interference of a particular laser frequency. They are typically accurate to <1% across the entire surface of the standard. These reference standards are remarkably durable under typical operating conditions. The surface contamination behavior is also very good. There are no better sub-micron reference standard available in this price range.
Mount: Pin
Spacing: 700 nm
Structure: Parallel Ridges
Material: W-coated Photoresist on Si
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.