Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences MXS Calibration Standard MXS 702CE for SEM, Pin 700 nm, 700 nm Spacing
SDP

Catalog No. 501901681
Encompass_Preferred

EMS offers a series of calibration standards with one and two dimension calibrated patterns. These standards are created utilizing holographic interference of a particular laser frequency. They are typically accurate to <1% across the entire surface of the standard. These reference standards are remarkably durable under typical operating conditions. The surface contamination behavior is also very good. There are no better sub-micron reference standard available in this price range.
Mount: Pin
Spacing: 700 nm
Structure: Arrayof Posts
Material: W-coated Photoresist on Si

Catalog No. 50-190-1681 Supplier Electron Microscopy Sciences Supplier No. 8011072PIN
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EMS offers a series of calibration standards with one and two dimension calibrated patterns. These standards are created utilizing holographic interference of a particular laser frequency. They are typically accurate to <1% across the entire surface of the standard. These reference standards are remarkably durable under typical operating conditions. The surface contamination behavior is also very good. There are no better sub-micron reference standard available in this price range.
Mount: Pin
Spacing: 700 nm
Structure: Arrayof Posts
Material: W-coated Photoresist on Si

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.