Learn More
Bachem FA-Phe-Gly-Gly-OH
SDP
Supplier:  Bachem 4003634.0250
Encompass_Preferred
FA-Phe-Gly-Gly-OH, 64967-39-1, C20H21N3O6, Mr 399.40, 250mg. FAPGG. Substrate for a continuous spectrophotometric assay of human angiotensin I-converting enzyme (ACE). Enzymatic cleavage of FAPGG yields FA-Phe-OH and H-Gly-Gly-OH.
Catalog No. 50-194-6743