Learn More
Bachem Z-Phe-Phe-OH
SDP
Supplier:  Bachem 4000601.0025
Encompass_Preferred
Z-Phe-Phe-OH, 13122-91-3, C26H26N2O5, Mr 446.50, 25g. Substrate for cathepsin A. As Fmoc-Phe-Phe-OH, Z-Phe-Phe-OH forms hydrogels.
Catalog No. 50-194-6998