Learn More
Bachem Bz-DL-Arg-bNA · HCl
SDP
Supplier:  Bachem 4000654.0005
Encompass_Preferred
Bz-DL-Arg-bNA · HCl, 913-04-2, C23H25N5O2 · HCl, Mr 439.95, 5g. DL-BANA. Substrate for cathepsin B, cathepsin H, and trypsin.
Catalog No. 50-194-7018