Learn More
Bachem H-Pro-Phe-OH
SDP
Supplier:  Bachem 4001974.0005
Encompass_Preferred
H-Pro-Phe-OH, 13589-02-1, C14H18N2O3, Mr 262.31, 5g. Substrate for human kidney prolinase (prolyl dipeptidase). ACE2 inhibitor, IC50 0.15 mM.
Catalog No. 50-194-6740