Learn More
Medchemexpress LLC Suc-AAPF-AMC (Suc-Ala-Ala-Pro-Phe-AMC) | 88467-45-2 | 99.5% | 661.70 | 1 ML

Supplier: Medchemexpress LLC HY13781110MM/1ML
Suc-AAPF-AMC is an alpha-chymotrypsin fluorogenic substrate for the quantitative determination of chymotrypsin activity. It is intended for research use only.
- Purity of 99.5%
- Supplied as a white to off-white solid
- Chemical formula C34H39N5O9
- Sequence: {Suc}-Ala-Ala-Pro-Phe-{AMC}
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.