Learn More
Cayman Chemical Suc-AAPF-pNA 100mg
SDP
Supplier:  Cayman Chemical 14993100 MG
Encompass_Preferred
A chromogenic substrate that can be cleaved by cathepsin G (Km = 1.7 mM), subtilisins, chymotrypsin (Km = 60 µM), chymase (Km = 4 mM), and cyclophilin; used for inhibitor screening and kinetic analysis
Catalog No. 50-297-2888