Learn More
Medchemexpress LLC Suc-AAPF-pNA (Suc-Ala-Ala-Pro-Phe-pNA) | 70967-97-4 | 99.4% | 624.64 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP25735MG
Encompass_Preferred
Suc-AAPF-pNA is a chromogenic p-nitroanilide (pNA) substrate with a Km of 1.7 mM. Its cleavage releases 4-nitroaniline, which is yellow and can be measured spectrophotometrically. This product can be used to measure free and membrane-bound cathepsin G in human neutrophils.
  • Chromogenic p-nitroanilide (pNA) substrate
  • Cleavage releases 4-nitroaniline, which is yellow
  • Can be measured spectrophotometrically
  • Suitable for measuring free and membrane-bound cathepsin G in human neutrophils
Catalog No. 50-004-24025