Learn More
Bachem Suc-Ala-Ala-Pro-Met-pNA
SDP
Supplier:  Bachem 4006760.0250
Encompass_Preferred
Suc-Ala-Ala-Pro-Met-pNA, 72682-73-6, C26H36N6O9S, Mr 608.67, 250mg. Suc-AAPM-pNA, a sensitive substrate for human pancreatic elastase.
Catalog No. 50-194-7062