Learn More
Carbosynth LLC. Suc-Ala-Ala-Ala-MCA (5 mg vial), 1ea, MW: 488.49, Molecular formula: C23H28N4O8, CAS# 73617-90-0
SDP
Supplier:  Carbosynth LLC. MAA3133V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Succinyl-L-Alanyl-L-Alanyl-L-Alanine 4-Methyl-Coumaryl-7-Amide, Suc-Ala-Ala-Ala-AMC, Suc-AAA-AMC, Suc-AAA-MCA, Application: Substrate for Elastase, Storage Temperature: -20C, References: R.A. Mumford, A.W. Strauss, J.C. Powers, P.A. Pierzchala, N. Nishino, and M. Zimmerman, J. Biol. Chem., 255, 2227 (1980).
Catalog No. 50-168-5925