Learn More
Medchemexpress LLC Suc-Ala-Ala-Phe-AMC | 71973-79-0 | MFCD00038448 | 99.9% | 564.59 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137337100MG
Encompass_Preferred
Suc-Ala-Ala-Phe-AMC is a fluorogenic chymotrypsin substrate that can be hydrolyzed by endopeptidase. It has been utilized in both in vivo assays of the acrosome reaction and in vitro enzyme assays.
  • Fluorogenic chymotrypsin substrate
  • Hydrolyzable by endopeptidase
  • Suitable for in vivo assays of acrosome reaction
  • Suitable for in vitro enzyme assays
  • Appears as a white to off-white solid
  • High purity of 99.89%
  • Soluble in DMSO at 250 mg/mL
Catalog No. 50-003-52154