Learn More
Carbosynth LLC. Suc-Ala-Ala-Pro-Phe-MCA (5 mg vial), 1ea, MW: 661.7, Molecular formula: C34H39N5O9, CAS# 88467-45-2
SDP
Supplier:  Carbosynth LLC. MAA3114V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Succinyl-L-Alanyl-L-Alanyl-L-Prolyl-L-Phenylalanine 4-Methyl-Coumaryl-7-Amide, Suc-Ala-Ala-Pro-Phe-AMC, Suc-AAPF-AMC, Suc-AAPF-MCA, Purity: , Grade A, Application: Substrate for Chymotrypsin, Storage Temperature: -20C, References: S. Sawada, H. Yokasawa, M. Hoshi and S. Ishii, Experientia, 39, 377 (1983).
Catalog No. 50-168-5924