Learn More
Medchemexpress LLC Suc-Ala-Glu-Pro-Phe-pNA | 128802-76-6 | 99.5% | 682.68 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP42025MG
Encompass_Preferred
Suc-Ala-Glu-Pro-Phe-pNA (Suc-AEPF-pNA) is a chromogenic substrate for the peptidyl-prolyl isomerase Pin1. It is utilized to assess the inhibitory effect of target compounds on Pin1, the catalytic activity of Pin1, and other related assays.
  • Functions as a substrate for recombinant human Pin1 in cell-free protease-coupled PPIase assays.
  • Enables measurement of Pin1 isomerase activity via pNA release.
  • Specifically elutes phage bound to the Pin1 PPIase domain active site.
  • Acts as a substrate for GST-tagged Pin1 PPIase activity.
  • Serves as a substrate for recombinant human Pin1's PPIase activity, which is inhibited by PKA phosphorylation.
Catalog No. 50-003-98198