Learn More
Bachem Suc-Ala-Glu-Pro-Phe-pNA
SDP
Supplier:  Bachem 4015607.0050
Encompass_Preferred
Suc-Ala-Glu-Pro-Phe-pNA, 128802-76-6, C32H38N6O11, Mr 682.69, 50mg. Suc-AEPF-pNA, chromogenic substrate for the peptidylprolyl isomerase Pin1.
Catalog No. 50-260-418