Learn More
Bachem Suc-Ala-Leu-Pro-Phe-pNA
SDP
Supplier:  Bachem 4015254.0050
Encompass_Preferred
Suc-Ala-Leu-Pro-Phe-pNA, 128802-78-8, C33H42N6O9, Mr 666.73, 50mg. Substrate for FK-506 binding proteins (FKBPs, also called macrophilins) and cyclophilins, which belong to the group of peptidyl prolyl cis-trans isomerases (PPIases). Suc-ALPF-pNA,has been used for an uncoupled protease-free assay of PPIase activity.
Catalog No. 50-260-417