Learn More
Medchemexpress LLC Suc-Ala-Phe-Lys-AMC | 73207-91-7 | 99.8% | 621.68 | 5 MG

Supplier: Medchemexpress LLC HYP43605MG
Suc-Ala-Phe-Lys-AMC is a highly sensitive fluorogenic substrate specifically designed for plasmin assays. It also serves as an effective substrate for assaying gingipain K, offering reliable performance for related research applications.
- Highly sensitive fluorogenic substrate for plasmin.
- Also used for assaying gingipain K.
- Purity of 99.8%.
- Stable for two years as powder at -80°C.
- Soluble in DMSO at 100 mg/mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.