Learn More
Medchemexpress LLC Suc-Gly-Pro-AMC | 80049-85-0 | 99.2% | C21H23N3O7 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY145983
Encompass_Preferred
Suc-Gly-Pro-AMC is a fluorescent substrate and a specific substrate for fibroblast activation protein (FAP), reacting with recombinant porcine prolyl oligopeptidase. It is used to study FAP and prolyl endopeptidase (PREP) activity in research, such as glioma studies. This solid product has a chemical formula of C21H23N3O7 and a purity of 99.16%. It is stable under recommended storage conditions but may emit toxic fumes if decomposed by fire. Not classified as hazardous, it is for research use only by qualified scientists in equipped facilities.
  • Appears as a solid, white to off-white
  • For use in laboratory chemical applications
  • Stable under recommended storage conditions
  • Store at 4°C, protected from light
  • Avoid strong acids/alkalis and strong oxidising/reducing agents
  • Not classified as a hazardous substance or mixture
Catalog No. 50-003-53077