Learn More
Medchemexpress LLC Suc-gly-pro-AMC | 80049-85-0 | MFCD00038445 | 99.2% | 429.42 g·mol⁻¹ | C21H23N3O7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1459835MG
Encompass_Preferred
Suc-Gly-Pro-AMC is a fluorogenic peptide substrate that releases 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage, enabling quantitative measurement of fibroblast activation protein and prolyl endopeptidase activity in biochemical assays. It is supplied as a high-purity solid suitable for enzyme kinetics, inhibitor screening, and related research applications.
  • High purity suitable for sensitive assays.
  • Releases AMC fluorescent reporter upon cleavage.
  • Suitable for fibroblast activation protein and prolyl endopeptidase assays.
  • Applicable to enzyme kinetics and inhibitor screening.
  • Recommended solvent: DMSO for stock solutions.
  • Store solid protected from light at 4°C; store solutions frozen as recommended.
Catalog No. 50-002-23001