Learn More
Medchemexpress LLC Suc-Ile-Glu(γ-pip)-Gly-Arg-pNA hydrochloride | 1379822-04-4 | 98.7% | 797.30 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP31261MG
Encompass_Preferred
Suc-Ile-Glu(γ-pip)-Gly-Arg-pNA hydrochloride is identified as a specific chromogenic substrate for factor Xa. It is intended for research use only and is not sold to patients. This product is shipped at room temperature within the continental US, with variations possible elsewhere. It requires sealed storage, away from moisture and light, and under nitrogen.
  • Factor Xa specific chromogenic substrate
  • High purity: 98.68%
  • Available in various quantities with bulk inquiry options
  • Soluble in DMSO (≥ 100 mg/mL)
  • Detailed handling and storage instructions provided
Catalog No. 50-003-96619